siRNA Details
Gene Symbol: Dnal4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017470.2 | NP_059498.2 | 3, 4 | 395 | ||
GenBank mRNA | AB010031.1 | - | 3, 4 | 373 | ||
AK029618.1 | - | 2, 3 | 206 | |||
AK132061.1 | - | 3, 4 | 385 | |||
BC005426.1 | - | 3, 4 | 324 | 3593932 |

More Information
Other Species Targeted

Gene Ontology Categories:
