siRNA Details
Gene Symbol: Kat6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205241.1 | NP_001192170.1 | 6, 7 | 1366 | ||
NM_017479.3 | NP_059507.2 | 6, 7 | 1490 | |||
GenBank mRNA | AF222800.1 | - | 6, 7 | 1406 | ||
AK045188.1 | - | 6, 7 | 1502 | |||
AK147509.1 | - | 6, 7 | 1367 | |||
BC095974.1 | - | 5, 6 | 1143 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
