siRNA Details
Gene Symbol: Rragc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017475.2 | NP_059503.2 | 2 | 522 | ||
GenBank mRNA | AB017616.1 | - | 2 | 445 | ||
AK150792.1 | - | 2 | 476 | |||
AK159355.1 | - | 2 | 455 | |||
AK159434.1 | - | 2 | 477 | |||
AK166658.1 | - | 2 | 468 | |||
BC005417.1 | - | 2 | 470 | 3495022 | ||
BC071245.1 | - | 2 | 481 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
