siRNA Details
Gene Symbol: Rragc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017475.2 | NP_059503.2 | 4 | 763 | ||
GenBank mRNA | AB017616.1 | - | 4 | 686 | ||
AK150792.1 | - | 4 | 717 | |||
AK159355.1 | - | 4 | 696 | |||
AK159434.1 | - | 4 | 718 | |||
AK166658.1 | - | 4 | 709 | |||
BC005417.1 | - | 4 | 711 | 3495022 | ||
BC071245.1 | - | 4 | 1111 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
