siRNA Details
Gene Symbol: Rabep1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291141.1 | NP_001278070.1 | 8 | 1361 | ||
NM_001291142.1 | NP_001278071.1 | 8 | 1370 | |||
NM_001291143.1 | NP_001278072.1 | 10 | 1561 | |||
NM_019400.3 | NP_062273.2 | 9 | 1490 | |||
GenBank mRNA | AB001750.1 | - | 10 | 1397 | ||
AF248489.1 | - | 8 | 1156 | |||
AF248490.1 | - | 8 | 1147 | |||
AK134587.1 | - | 9 | 1491 | |||
AK137236.1 | - | 9 | 1455 | |||
BC003921.1 | - | 1 | 216 | |||
BC060166.1 | - | 9 | 1508 | |||
BC080274.1 | - | 9 | 1462 | |||
BC129853.1 | - | 9 | 1309 | 40109688 | ||
BC129854.1 | - | 9 | 1309 | 40109689 | ||
D86066.1 | - | 9 | 1326 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
