siRNA Details
Gene Symbol: Pabpn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019402.2 | NP_062275.1 | 4 | 606 | ||
GenBank mRNA | AK032172.1 | - | 4 | 602 | ||
AK157143.1 | - | 4 | 595 | |||
BC049869.1 | - | 4 | 591 | |||
BC055866.1 | - | 4 | 595 | 4218469 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
