siRNA Details
Gene Symbol: Rnf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019403.3 | NP_062276.1 | 5 | 500 | ||
GenBank mRNA | AK010792.1 | - | 5 | 530 | ||
AK012729.1 | - | 5 | 492 | |||
AK019289.1 | - | 5 | 511 | |||
AK020514.2 | - | 5 | 501 | |||
AK020515.2 | - | 5 | 519 | |||
BC012031.1 | - | 5 | 452 | |||
BC016449.1 | - | 5 | 500 | 4191746 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
