siRNA Details
Gene Symbol: Rnf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019403.3 | NP_062276.1 | 1 | 165 | ||
GenBank mRNA | AK012729.1 | - | 1 | 157 | ||
AK019289.1 | - | 1 | 176 | |||
AK020514.2 | - | 1 | 166 | |||
AK020515.2 | - | 1 | 184 | |||
BC012031.1 | - | 1 | 116 | |||
BC016449.1 | - | 1 | 165 | 4191746 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
