siRNA Details
Gene Symbol: Ccrl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB009384.1 | - | 3 | 449 | ||
AF030185.1 | - | 2 | 625 | |||
AF316576.1 | - | 2 | 400 | |||
AJ318863.1 | - | 2 | 239 | |||
AK007808.1 | - | 2 | 698 | |||
AK145946.1 | - | 2 | 673 | |||
AK151856.1 | - | 3 | 449 | |||
BC038631.1 | - | 3 | 358 | 1347006 | ||
CT010248.1 | - | 1 | 181 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
