siRNA Details
Gene Symbol: Zfp316 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017467.3 | NP_059495.3 | 7 | 1646 | ||
GenBank mRNA | AF082568.1 | - | 1 | 1259 | ||
BC057078.1 | - | 7 | 1563 | 5698016 | ||
BC060099.1 | - | 5 | 1168 |

More Information
Other Species Targeted

Gene Ontology Categories:
