siRNA Details
Gene Symbol: Zfp316 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017467.3 | NP_059495.3 | 4 | 713 | ||
GenBank mRNA | BC057078.1 | - | 4 | 630 | 5698016 | |
BC060099.1 | - | 2 | 236 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
