siRNA Details
Gene Symbol: Sept1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017461.2 | NP_059489.2 | 5 | 381 | ||
GenBank mRNA | BB610735.1 | - | 5 | 381 | ||
BC140998.1 | - | 5 | 382 | 9055610 | ||
BC145081.1 | - | 5 | 382 | 9053629 | ||
M37030.1 | - | 5 | 388 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
