siRNA Details
Gene Symbol: Elovl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039175.2 | NP_001034264.1 | 6 | 491 | ||
NM_001039176.2 | NP_001034265.1 | 7 | 689 | |||
NM_019422.3 | NP_062295.1 | 7 | 682 | |||
GenBank mRNA | AF104033.1 | - | 4 | 248 | ||
AF170907.1 | - | 7 | 675 | |||
AK003743.1 | - | 6 | 460 | |||
AK135348.1 | - | 7 | 657 | |||
AK156003.1 | - | 7 | 2283 | |||
AK171217.1 | - | 7 | 668 | |||
AK195478.1 | - | 2 | 156 | |||
BC006735.1 | - | 7 | 689 | 3156004 | ||
BC096673.1 | - | 6 | 470 | 5143739 | ||
CN455861.1 | - | 5 | 426 | |||
CX199666.1 | - | 7 | 655 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
