siRNA Details
Gene Symbol: Elp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253700.1 | NP_001240629.1 | 6 | 566 | ||
NM_018740.2 | NP_061210.2 | 8 | 1048 | |||
GenBank mRNA | AF004107.1 | - | 7 | 967 | ||
AK011130.1 | - | 6 | 860 | |||
AK152582.1 | - | 8 | 1052 | |||
BC003752.1 | - | 7 | 899 | 3489878 | ||
BE862527.1 | - | 3 | 324 |

More Information
Other Species Targeted

Gene Ontology Categories:
