siRNA Details
Gene Symbol: Epb4.1l4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019427.2 | NP_062300.2 | 12 | 1209 | ||
GenBank mRNA | AB032366.1 | - | 12 | 1704 | ||
AK144315.1 | - | 12 | 1295 | |||
AK165783.1 | - | 12 | 1202 | |||
BC113123.1 | - | 13 | 1031 | |||
CF915899.1 | - | 2 | 162 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
