siRNA Details
Gene Symbol: Epb4.1l4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019427.2 | NP_062300.2 | 4 | 480 | ||
GenBank mRNA | AB032366.1 | - | 4 | 975 | ||
AK144315.1 | - | 4 | 566 | |||
AK165783.1 | - | 4 | 473 | |||
BB612044.1 | - | 4 | 481 | |||
BC113123.1 | - | 5 | 324 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
