siRNA Details
Gene Symbol: Prss16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019429.2 | NP_062302.1 | 9 | 1076 | ||
GenBank mRNA | AJ131775.1 | - | 9 | 1076 | ||
AK037514.1 | - | 5 | 1895 | |||
AK040376.1 | - | 2 | 459 | |||
AK088019.1 | - | 9 | 1104 | |||
BC125481.1 | - | 9 | 1074 | 40129996 | ||
BC131955.1 | - | 9 | 1074 | 40130232 |

More Information
Other Species Targeted

Gene Ontology Categories:
