siRNA Details
Gene Symbol: Cacng3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019430.2 | NP_062303.2 | 1 | 88 | ||
GenBank mRNA | AJ272044.1 | - | 1 | 58 | ||
BC138673.1 | - | 1 | 88 | 8861694 | ||
BC145971.1 | - | 1 | 88 | 40131274 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
