siRNA Details
Gene Symbol: Cacng4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019431.2 | NP_062304.1 | 4 | 511 | ||
GenBank mRNA | AJ272045.1 | - | 4 | 511 | ||
AK144253.1 | - | 4 | 742 | |||
BC138503.1 | - | 4 | 530 | 8861523 | ||
BC138506.1 | - | 4 | 530 | 8861526 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
