siRNA Details
Gene Symbol: Cacng6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133183.1 | NP_573446.1 | 2 | 344 | ||
GenBank mRNA | AF361348.1 | - | 2 | 344 | ||
BC115456.1 | - | 2 | 344 | 40100157 | ||
BC115457.1 | - | 2 | 344 | 40100159 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
