siRNA Details
Gene Symbol: Tcstv1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018756.3 | NP_061226.1 | 2 | 404 | ||
GenBank mRNA | AF067057.1 | - | 2 | 386 | ||
AF067059.1 | - | 2 | 404 | |||
BC119476.2 | - | 1 | 888 | 40049850 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
