siRNA Details
Gene Symbol: Rfk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019437.3 | NP_062310.1 | 4 | 675 | ||
GenBank mRNA | AK002806.1 | - | 4 | 636 | ||
AK008352.1 | - | 4 | 605 | |||
AK010607.1 | - | 4 | 604 | |||
BC033521.1 | - | 4 | 577 | 4527980 | ||
BC051021.1 | - | 4 | 586 | 6315425 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
