siRNA Details
Gene Symbol: Ncapg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019438.1 | NP_062311.1 | 19, 20 | 2920 | ||
GenBank mRNA | AJ237585.1 | - | 12, 13 | 3607 | ||
AK136254.1 | - | 14, 15 | 1906 | |||
BC137834.1 | - | 12, 13 | 2138 | 8860853 | ||
BC144960.1 | - | 12, 13 | 2138 | 9053508 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
