siRNA Details
Gene Symbol: Stk19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019442.3 | NP_062315.1 | 3 | 295 | ||
GenBank mRNA | AF099934.1 | - | 3 | 229 | ||
AK146739.1 | - | 3 | 297 | |||
AK168865.1 | - | 3 | 297 | |||
AK215314.1 | - | 2 | 55 | |||
AK218509.1 | - | 2 | 124 | |||
BC022681.1 | - | 3 | 295 | 4240347 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
