siRNA Details
Gene Symbol: Slc4a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136260.1 | NP_001129732.1 | 12 | 1418 | ||
NM_001197147.1 | NP_001184076.1 | 12 | 1391 | |||
NM_018760.2 | NP_061230.2 | 12 | 1418 | |||
GenBank mRNA | AF141934.3 | - | 9 | 1220 | ||
BC026592.1 | - | 3 | 211 | |||
BC129863.1 | - | 12 | 1380 | |||
BC129864.1 | - | 12 | 1379 | |||
BC148292.2 | - | 12 | 1354 | 40110074 | ||
HQ018820.1 | - | 9 | 1255 | |||
HQ204220.1 | - | 12 | 1362 | |||
HQ285250.1 | - | 9 | 1231 |

More Information
Other Species Targeted

Gene Ontology Categories:
