siRNA Details
Gene Symbol: Slc4a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136260.1 | NP_001129732.1 | 6 | 775 | ||
NM_018760.2 | NP_061230.2 | 6 | 775 | |||
GenBank mRNA | AF020195.1 | - | 6 | 738 | ||
AF141934.3 | - | 3 | 577 | |||
AK140443.1 | - | 6 | 1218 | |||
BC129863.1 | - | 6 | 737 | |||
BC129864.1 | - | 6 | 736 | |||
HQ018820.1 | - | 3 | 612 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
