siRNA Details
Gene Symbol: Nfat5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286260.1 | NP_001273189.1 | 3 | 959 | ||
NM_018823.2 | NP_061293.2 | 4 | 1013 | |||
NM_133957.3 | NP_598718.2 | 5 | 1075 | |||
GenBank mRNA | AF162853.1 | - | 3 | 870 | ||
AF200687.1 | - | 4 | 648 | |||
AF228705.1 | - | 1 | 713 | |||
AF228706.1 | - | 1 | 713 | |||
AF369980.1 | - | 5 | 714 | |||
AF453571.2 | - | 5 | 683 | |||
AK134022.1 | - | 5 | 1079 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
