siRNA Details
Gene Symbol: Nfat5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286260.1 | NP_001273189.1 | 3 | 953 | ||
NM_018823.2 | NP_061293.2 | 4 | 1007 | |||
NM_133957.3 | NP_598718.2 | 5 | 1069 | |||
GenBank mRNA | AF162853.1 | - | 3 | 864 | ||
AF200687.1 | - | 4 | 642 | |||
AF228705.1 | - | 1 | 707 | |||
AF228706.1 | - | 1 | 707 | |||
AF369980.1 | - | 5 | 708 | |||
AF453571.2 | - | 5 | 677 | |||
AK134022.1 | - | 5 | 1073 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001107425, XM_006255553, XM_008772504, XM_008772503, XM_003751872.
Gene Ontology Categories:
