siRNA Details
Gene Symbol: Il1f6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019450.3 | NP_062323.1 | 4, 5 | 440 | ||
GenBank mRNA | AF200493.1 | - | 3, 4 | 279 | ||
AF206697.1 | - | 4, 5 | 354 | |||
AK004061.1 | - | 4, 5 | 440 | |||
AK224931.1 | - | 4, 5 | 374 | |||
AK224932.1 | - | 4, 5 | 387 | |||
BC117102.1 | - | 3, 4 | 313 | 40126421 | ||
BC117104.1 | - | 3, 4 | 313 | 40126423 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
