siRNA Details
Gene Symbol: Tbc1d8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018775.4 | NP_061245.3 | 12, 13 | 2360 | ||
GenBank mRNA | AK138630.1 | - | 12, 13 | 2358 | ||
AK141520.1 | - | 12, 13 | 2349 | |||
AK141816.1 | - | 12, 13 | 2314 | |||
AK162371.1 | - | 6, 7 | 1135 | |||
BC005421.1 | - | 9, 10 | 1695 | |||
BC065081.1 | - | 12, 13 | 2294 | 6832495 | ||
U88873.1 | - | 8, 9 | 1580 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001191067, XM_006244851.
Gene Ontology Categories:
