siRNA Details
Gene Symbol: Wdr45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290792.1 | NP_001277721.1 | 8 | 835 | ||
NM_001290794.1 | NP_001277723.1 | 8 | 731 | |||
NM_001290795.1 | NP_001277724.1 | 8 | 835 | |||
NM_172372.2 | NP_758960.1 | 8 | 831 | |||
GenBank mRNA | AF229635.1 | - | 8 | 809 | ||
AK145616.1 | - | 8 | 731 | |||
AK205782.1 | - | 3 | 297 | |||
AK208276.1 | - | 3 | 296 | |||
BC011479.1 | - | 8 | 802 | 4035468 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
