siRNA Details
Gene Symbol: Otud5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290536.1 | NP_001277465.1 | 5 | 1272 | ||
NM_001290537.1 | NP_001277466.1 | 6 | 892 | |||
NM_138604.3 | NP_613070.2 | 5 | 1272 | |||
GenBank mRNA | AF229642.1 | - | 5 | 413 | ||
AK149777.1 | - | 6 | 893 | |||
AK155131.1 | - | 5 | 1248 | |||
AK167226.1 | - | 4 | 585 | |||
AK169921.1 | - | 7 | 858 | |||
AK171874.1 | - | 5 | 1013 | |||
AK179391.1 | - | 11 | ||||
BC016529.1 | - | 5 | 1272 | 3594583 | ||
BC051111.1 | - | 5 | 1011 | 5253679 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001037496, XM_006256741, XM_006256737, XM_006256738, XM_006256739, and others.
Gene Ontology Categories:
