siRNA Details
Gene Symbol: Sfmbt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166531.1 | NP_001160003.1 | 9, 10 | 1172 | ||
NM_001166532.1 | NP_001160004.1 | 8, 9 | 1266 | |||
NM_019460.2 | NP_062333.1 | 8, 9 | 1123 | |||
GenBank mRNA | AB032165.1 | - | 7, 8 | 968 | ||
AK142330.1 | - | 8, 9 | 1150 | |||
AK154826.1 | - | 8, 9 | 1268 | |||
AK160573.1 | - | 9, 10 | 1172 | |||
BC018217.1 | - | 8, 9 | 1134 | |||
BC036972.1 | - | 8, 9 | 1193 | |||
BC066188.1 | - | 8, 9 | 1076 | 30476111 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
