siRNA Details
Gene Symbol: Atp8b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081182.2 | NP_001074651.2 | 25 | 3015 | ||
GenBank mRNA | AF156546.1 | - | 3 | 274 | ||
AK155736.1 | - | 24 | 3049 | |||
AK173107.1 | - | 14 | 2460 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
