siRNA Details
Gene Symbol: KRT16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF061812.1 | - | 2, 3 | 631 | ||
AK290853.1 | - | 2, 3 | 694 | |||
S79867.1 | - | 2, 3 | 617 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
