siRNA Details
Gene Symbol: Eef1b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018796.3 | NP_061266.2 | 2 | 546 | ||
GenBank mRNA | AF029844.2 | - | 2 | 212 | ||
AK012756.1 | - | 2 | 249 | |||
AK027911.1 | - | 2 | 249 | |||
AK168191.1 | - | 2 | 248 | |||
BC003899.1 | - | 2 | 242 | 3600850 | ||
BC023139.1 | - | 2 | 258 | 3963930 | ||
BC039635.1 | - | 2 | 261 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
