siRNA Details
Gene Symbol: Mpc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018819.4 | NP_061289.1 | 4 | 340 | ||
GenBank mRNA | AF181116.1 | - | 4 | 296 | ||
AK002241.1 | - | 4 | 341 | |||
AK002266.1 | - | 4 | 319 | |||
AK004013.1 | - | 4 | 296 | |||
AK005348.1 | - | 4 | 332 | |||
AK013391.1 | - | 4 | 341 | |||
AK014421.1 | - | 4 | 298 | |||
AK014435.1 | - | 4 | 570 | |||
AK135943.1 | - | 4 | 311 | |||
AK170514.1 | - | 4 | 357 | |||
BC024365.1 | - | 4 | 302 | 4988147 | ||
BC103767.1 | - | 4 | 343 | 30840199 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
