siRNA Details
Gene Symbol: Camkk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018883.2 | NP_061371.2 | 4, 5 | 622 | ||
GenBank mRNA | AF117384.1 | - | 4, 5 | 577 | ||
AK144483.1 | - | 4, 5 | 623 | |||
BC017529.1 | - | 4, 5 | 587 | 4924656 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_031662, XM_006246801, XM_008767866, XM_006246799, L42810, and others.
Gene Ontology Categories:
