siRNA Details
Gene Symbol: Cxcl13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018866.2 | NP_061354.1 | 2 | 175 | ||
GenBank mRNA | AF030636.1 | - | 2 | 171 | ||
AF044196.1 | - | 2 | 175 | |||
AK156025.1 | - | 2 | 189 | |||
AK160241.1 | - | 2 | 191 | |||
BB612247.1 | - | 186 | ||||
BC012965.1 | - | 2 | 174 | 4217539 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
