siRNA Details
Gene Symbol: Trim3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285870.1 | NP_001272799.1 | 10 | 2142 | ||
NM_001285871.1 | NP_001272800.1 | 10 | 2327 | |||
NM_001285873.1 | NP_001272802.1 | 10 | 2142 | |||
NM_018880.3 | NP_061368.1 | 10 | 2116 | |||
GenBank mRNA | AB030912.1 | - | 10 | 2143 | ||
AF220019.1 | - | 10 | 2035 | |||
AK019165.1 | - | 10 | 2077 | |||
AK144674.1 | - | 10 | 2045 | |||
AK170029.1 | - | 10 | 2080 | |||
AK182206.1 | - | 1 | 55 | |||
BC003347.1 | - | 10 | 2156 | |||
BC034263.1 | - | 10 | 2081 | 4503884 |

More Information
Other Species Targeted

Gene Ontology Categories:
