siRNA Details
Gene Symbol: Smad9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019483.5 | NP_062356.3 | 3 | 969 | ||
GenBank mRNA | AK137411.1 | - | 3 | 970 | ||
AY145520.1 | - | 2 | 802 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
