siRNA Details
Gene Symbol: Nprl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF131207.1 | - | 5 | 730 | ||
AK088925.1 | - | 5 | 696 | |||
BC026548.1 | - | 5 | 647 | 5028027 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
