siRNA Details
Gene Symbol: Gtpbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145979.1 | NP_001139451.1 | 4 | 552 | ||
NM_019581.3 | NP_062527.2 | 4 | 587 | |||
GenBank mRNA | AB024573.1 | - | 2 | 223 | ||
AF168991.1 | - | 4 | 492 | |||
AK087511.1 | - | 4 | 458 | |||
AK146415.1 | - | 4 | 537 | |||
AK148020.1 | - | 4 | 508 | |||
BC049089.1 | - | 4 | 548 | 5720710 | ||
BC051958.1 | - | 4 | 548 | 5720710 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
