siRNA Details
Gene Symbol: Il17b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019508.1 | NP_062381.1 | 2 | 322 | ||
GenBank mRNA | AF184970.1 | - | 2 | 322 | ||
AF218726.1 | - | 2 | 322 | |||
AF250308.1 | - | 2 | 319 | |||
AK003506.1 | - | 2 | 380 | |||
BC002271.1 | - | 2 | 322 | 3496106 |
