siRNA Details
Gene Symbol: Lgals12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019516.3 | NP_062389.1 | 4 | 891 | ||
GenBank mRNA | AF223223.1 | - | 5 | 585 | ||
AF244978.1 | - | 5 | 593 | |||
AF244979.1 | - | 4 | 638 | |||
AK033535.1 | - | 4 | 818 | |||
AK085660.1 | - | 4 | 894 | |||
AK136526.1 | - | 4 | 892 | |||
BC030890.1 | - | 4 | 575 | 4485119 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
