siRNA Details
Gene Symbol: Astn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019514.3 | NP_062387.2 | 1 | 556 | ||
NM_207109.2 | NP_996992.1 | 1 | 556 | |||
GenBank mRNA | AK041160.1 | - | 2 | 460 | ||
AK047058.1 | - | 1 | 545 | |||
AK051472.1 | - | 1 | 557 | |||
AK137377.1 | - | 1 | 547 |

More Information
Other Species Targeted

Gene Ontology Categories:
