siRNA Details
Gene Symbol: Ubqln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026842.4 | NP_081118.4 | 9 | 1556 | ||
NM_152234.2 | NP_689420.1 | 8 | 1472 | |||
GenBank mRNA | AK004183.1 | - | 2 | 840 | ||
AK048534.1 | - | 8 | 1393 | |||
AK049298.1 | - | 9 | 1311 | |||
AK131766.1 | - | 2 | 168 | |||
AK165456.1 | - | 8 | 1437 | |||
AK172695.1 | - | 8 | 1441 | |||
AK181884.1 | - | 2 | 86 | |||
AK186933.1 | - | 2 | 86 | |||
AK187729.1 | - | 3 | 170 | |||
AK198908.1 | - | 3 | 170 | |||
AK209853.1 | - | 3 | 170 | |||
BC010213.1 | - | 6 | 899 | |||
BC026847.1 | - | 9 | 1539 | 4015641 | ||
BC027375.1 | - | 3 | 259 | |||
BC028857.1 | - | 8 | 1411 | 4507759 | ||
BC051098.1 | - | 8 | 1184 | |||
BC080667.1 | - | 9 | 1517 | 4935292 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
