siRNA Details
Gene Symbol: Cts7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019539.3 | NP_062412.1 | 5 | 661 | ||
GenBank mRNA | AF250837.1 | - | 4 | 448 | ||
AY014779.1 | - | 5 | 521 | |||
BC050198.1 | - | 4 | 757 | |||
BC064740.1 | - | 5 | 661 | 30018572 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
