siRNA Details
Gene Symbol: Cts8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019541.3 | NP_062414.3 | 7 | 914 | ||
GenBank mRNA | AF250840.1 | - | 7 | 926 | ||
AK049474.1 | - | 7 | 915 | |||
AY014780.1 | - | 7 | 913 | |||
BC068241.1 | - | 7 | 904 | 30019120 |

More Information
Other Species Targeted

Gene Ontology Categories:
