siRNA Details
Gene Symbol: Plac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019538.4 | NP_062411.1 | 3 | 462 | ||
GenBank mRNA | AF234653.1 | - | 3 | 463 | ||
AF250838.1 | - | 3 | 459 | |||
AK005501.1 | - | 3 | 463 | |||
AK145876.1 | - | 3 | 428 | |||
BC051666.1 | - | 3 | 448 | 30017089 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
